siRNA Details
Gene Symbol: SMG1P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002453.5 | 6 | 1015 | |||
GenBank mRNA | BC063703.1 | - | 6 | 1015 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SMG1P5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002453.5 | 6 | 1015 | |||
GenBank mRNA | BC063703.1 | - | 6 | 1015 |