siRNA Details
Gene Symbol: LINC01300 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024441.1 | 4 | 667 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE535333.1 | - | 3 | 539 | Chr. 8 - 141342345 - 141344611 [+] on Build GRCh38 |
