siRNA Details
Gene Symbol: PIN1P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023916.1 | 1 | 352 | |||
GenBank mRNA | U82382.1 | - | 1 | 352 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR748481.1 | - | 1 | 305 | Chr. 1 - 69919369 - 69920015 [+] on Build GRCh38 |
