siRNA Details
Gene Symbol: LINC00312 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024065.2 | 1 | 512 | |||
GenBank mRNA | AF086709.2 | - | 1 | 89 | ||
AK054898.1 | - | 1 | 512 | |||
BC016278.1 | - | 1 | 210 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA283550.1 | - | 1 | 206 | Chr. 3 - 8572088 - 8572650 [+] on Build GRCh38 |
