siRNA Details
Gene Symbol: CNTFR-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024369.1 | 2 | 411 | |||
GenBank mRNA | BC033987.1 | - | 3 | 521 | ||
BC038740.1 | - | 5 | 843 | |||
BC042806.1 | - | 2 | 411 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI460276.1 | - | 2 | 415 | Chr. 9 - 34568012 - 34582829 [+] on Build GRCh38 |
