siRNA Details
Gene Symbol: FAM83A-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024479.1 | 1 | 16 | |||
GenBank mRNA | AY211906.1 | - | 1 | 16 | ||
AY211907.1 | - | 1 | 16 | |||
AY211908.1 | - | 1 | 16 | |||
AY225521.1 | - | 1 | 16 |
