siRNA Details
Gene Symbol: LOC284023 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024349.1 | 2 | 1060 | |||
GenBank mRNA | AK091084.1 | - | 1 | 875 | ||
AK095766.1 | - | 2 | 1054 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC284023 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024349.1 | 2 | 1060 | |||
GenBank mRNA | AK091084.1 | - | 1 | 875 | ||
AK095766.1 | - | 2 | 1054 |