siRNA Details
Gene Symbol: PCDHB18P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_001281.2 | 1 | 2705 | |||
GenBank mRNA | AF217743.1 | - | 1 | 2324 | ||
AK095496.1 | - | 1 | 2674 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE326289.1 | - | 1 | 16 | Chr. 5 - 141237022 - 141237440 [+] on Build GRCh38 |
