siRNA Details
Gene Symbol: MIR181B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029612.1 | 1 | 90 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NR_029820, AK053260, AK053260.
