siRNA Details
Gene Symbol: MIR196A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029617.1 | 1 | 25 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NR_029722.
