siRNA Details
Gene Symbol: LOC101928725 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135474.1 | 2 | 1173 | |||
GenBank mRNA | BC034806.1 | - | 2 | 1173 |
Gene Symbol: LOC100288175 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC033949.2 | - | 4 | 928 |
