siRNA Details
Gene Symbol: LINC00696 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027331.1 | 1 | 1763 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK127958.1 | - | 1 | 1763 | Chr. 3 - 52062094 - 52065112 [-] on Build GRCh38 |
