siRNA Details
Gene Symbol: LOC374443 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002814.2 | 1 | 660 | |||
NR_046444.1 | 1 | 660 | ||||
GenBank mRNA | AK096274.1 | - | 1 | 662 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC374443 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002814.2 | 1 | 660 | |||
NR_046444.1 | 1 | 660 | ||||
GenBank mRNA | AK096274.1 | - | 1 | 662 |