siRNA Details
Gene Symbol: LOC374443 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002814.2 | 4 | 1442 | |||
NR_046444.1 | 5 | 1525 | ||||
NR_046445.1 | 5 | 967 | ||||
NR_046446.1 | 5 | 919 | ||||
NR_046447.1 | 4 | 837 | ||||
NR_046448.1 | 5 | 837 | ||||
NR_046449.2 | 4 | 791 | ||||
NR_046450.1 | 5 | 705 | ||||
NR_046451.1 | 4 | 626 | ||||
GenBank mRNA | AK096274.1 | - | 4 | 1444 | ||
BF128895.1 | - | 5 | 690 | |||
BG754872.1 | - | 5 | 706 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX349220.1 | - | 4 | 718 | Chr. 12 - 9648781 - 9657434 [+] on Build GRCh38 |
