siRNA Details
Gene Symbol: LOC374443 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002814.2 | 4 | 1505 | |||
NR_046444.1 | 5 | 1588 | ||||
NR_046445.1 | 5 | 1030 | ||||
NR_046446.1 | 5 | 982 | ||||
NR_046447.1 | 4 | 900 | ||||
NR_046448.1 | 5 | 900 | ||||
NR_046449.2 | 4 | 854 | ||||
NR_046450.1 | 5 | 768 | ||||
NR_046451.1 | 4 | 689 | ||||
GenBank mRNA | AK096274.1 | - | 4 | 1507 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX349220.1 | - | 4 | 782 | Chr. 12 - 9648781 - 9657434 [+] on Build GRCh38 |
