siRNA Details
Gene Symbol: LOC374443 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002814.2 | 4 | 2027 | |||
NR_046444.1 | 5 | 2110 | ||||
NR_046445.1 | 5 | 1552 | ||||
NR_046446.1 | 5 | 1504 | ||||
NR_046447.1 | 4 | 1422 | ||||
NR_046448.1 | 5 | 1422 | ||||
NR_046449.2 | 4 | 1376 | ||||
NR_046450.1 | 5 | 1290 | ||||
NR_046451.1 | 4 | 1211 | ||||
GenBank mRNA | AK096274.1 | - | 4 | 2028 |
