siRNA Details
Gene Symbol: LOC374443 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002814.2 | 4 | 1956 | |||
NR_046444.1 | 5 | 2039 | ||||
NR_046445.1 | 5 | 1481 | ||||
NR_046446.1 | 5 | 1433 | ||||
NR_046447.1 | 4 | 1351 | ||||
NR_046448.1 | 5 | 1351 | ||||
NR_046449.2 | 4 | 1305 | ||||
NR_046450.1 | 5 | 1219 | ||||
NR_046451.1 | 4 | 1140 | ||||
GenBank mRNA | AK096274.1 | - | 4 | 1957 |
