siRNA Details
Gene Symbol: LINC00483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073199.1 | 3, 4 | 360 | |||
GenBank mRNA | AK000670.1 | - | 3, 4 | 361 | ||
AK000701.1 | - | 2, 3 | 2295 | |||
BP382918.1 | - | 3, 4 | 360 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG959766.1 | - | 399 | Chr. - - [-] on Build |
