siRNA Details
Gene Symbol: LINC00483 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073199.1 | 2 | 100 | |||
GenBank mRNA | AK000670.1 | - | 2 | 101 | ||
AK000701.1 | - | 1 | 556 | |||
AK222717.1 | - | 1 | 554 | |||
AK310037.1 | - | 2 | 142 | |||
BC128165.1 | - | 1 | 455 | |||
BC128166.1 | - | 1 | 455 | |||
BP382918.1 | - | 2 | 100 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA925984.1 | - | 2 | 142 | Chr. 17 - 50763215 - 50767557 [-] on Build GRCh38 |
