siRNA Details
Gene Symbol: LOC284412 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029390.1 | 1 | 1347 | |||
GenBank mRNA | AF289566.1 | - | 1 | 586 | ||
BC045185.1 | - | 1 | 1354 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC284412 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029390.1 | 1 | 1347 | |||
GenBank mRNA | AF289566.1 | - | 1 | 586 | ||
BC045185.1 | - | 1 | 1354 |