siRNA Details
Gene Symbol: LINC00663 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026956.1 | 3 | 1182 | |||
GenBank mRNA | AK022418.1 | - | 1 | 816 | ||
AK289657.1 | - | 3 | 1162 | |||
BC034236.1 | - | 4 | 1314 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00663 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026956.1 | 3 | 1182 | |||
GenBank mRNA | AK022418.1 | - | 1 | 816 | ||
AK289657.1 | - | 3 | 1162 | |||
BC034236.1 | - | 4 | 1314 |