siRNA Details
Gene Symbol: ZNF571 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK131351.1 | - | 1 | 3176 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK125901.1 | - | 1 | 4923 | Chr. 3 - 45681193 - 45686338 [+] on Build GRCh38 |
