siRNA Details
Gene Symbol: LOC100128554 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_130748.1 | 8, 9 | 1595 | |||
GenBank mRNA | BC035235.1 | - | 9, 10 | 1771 | ||
BC039370.1 | - | 8, 9 | 1595 | |||
BC118583.1 | - | 8, 9 | 1139 | 40085030 | ||
BC121801.1 | - | 9, 10 | 1378 |
