siRNA Details
Gene Symbol: LINC00239 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026774.1 | 4 | 425 | |||
GenBank mRNA | BX113211.1 | - | 4 | 425 | ||
CK904714.1 | - | 2 | 167 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC00239 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026774.1 | 4 | 425 | |||
GenBank mRNA | BX113211.1 | - | 4 | 425 | ||
CK904714.1 | - | 2 | 167 |