siRNA Details
Gene Symbol: LOC440173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027471.1 | 2 | 1933 | |||
GenBank mRNA | BC027471.1 | - | 2 | 1931 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BE736107.1 | - | 1 | 308 | Chr. 9 - 87008453 - 87008841 [-] on Build GRCh38 |
