siRNA Details
Gene Symbol: LINC00303 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027902.1 | 5 | 656 | |||
GenBank mRNA | AK097662.1 | - | 4 | 1033 | ||
BC104010.2 | - | 4 | 745 | 40024418 | ||
BC104011.2 | - | 5 | 656 | 40024419 | ||
BC104012.2 | - | 4 | 745 | 40024422 |
