siRNA Details
Gene Symbol: LOC284661 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027088.1 | 1 | 264 | |||
GenBank mRNA | AK093498.1 | - | 1 | 264 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB043752.1 | - | 1 | 155 | Chr. 1 - 4412161 - 4422877 [+] on Build GRCh38 |
