siRNA Details
Gene Symbol: CBLL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024199.2 | 3 | 663 | |||
XM_011516580.2 | XP_011514882.1 | 3 | 863 | |||
XM_017012645.1 | XP_016868134.1 | 3 | 715 | |||
GenBank mRNA | BP201841.1 | - | 3 | 215 | ||
DB033469.1 | - | 3 | 218 | |||
DB174309.1 | - | 3 | 525 |
