siRNA Details
Gene Symbol: LINC00942 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028415.1 | 2 | 622 | |||
GenBank mRNA | AK090616.1 | - | 2 | 594 | ||
BQ942184.1 | - | 2 | 532 | |||
BQ949914.1 | - | 1 | 115 | |||
BQ954540.1 | - | 2 | 551 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CD172419.1 | - | 2 | 572 | Chr. 12 - 1500545 - 1502837 [+] on Build GRCh38 |
