siRNA Details
Gene Symbol: PPP4R1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003505.3 | 2, 3 | 58 | |||
GenBank mRNA | AK057583.1 | - | 2, 3 | 58 | ||
AK299330.1 | - | 1, 2 | 52 | |||
BC100979.1 | - | 2, 3 | 58 | |||
BC100980.1 | - | 2, 3 | 58 |
