siRNA Details
Gene Symbol: PPP4R1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003505.3 | 14 | 1964 | |||
GenBank mRNA | AK057583.1 | - | 14 | 1964 | ||
CR994964.1 | - | 4 | 414 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CD654184.1 | - | 3 | 308 | Chr. 20 - 58232793 - 58239773 [-] on Build GRCh38 |
