siRNA Details
Gene Symbol: PPP4R1L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003505.3 | 16 | 2178 | |||
GenBank mRNA | CR994964.1 | - | 6 | 628 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CD654184.1 | - | 5 | 522 | Chr. 20 - 58232793 - 58239773 [-] on Build GRCh38 |
