siRNA Details
Gene Symbol: GSTTP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003082.1 | 1 | 1144 | |||
GenBank mRNA | AK127991.1 | - | 1 | 1144 |
Gene Symbol: GSTTP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC098108.1 | - | 2 | 130 |
