siRNA Details
Gene Symbol: CD209 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026692.1 | 1 | 194 | |||
GenBank mRNA | AY042229.1 | - | 1 | 127 | ||
AY042230.1 | - | 1 | 127 | |||
AY042231.1 | - | 1 | 127 | |||
AY042232.1 | - | 1 | 127 | |||
AY042233.1 | - | 1 | 127 | |||
BC110615.2 | - | 1 | 134 | 40032459 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX472296.1 | - | 1 | 225 | Chr. 19 - 7745877 - 7747605 [-] on Build GRCh38 |
