siRNA Details
Gene Symbol: FAM182A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026713.1 | 5 | 2242 | |||
GenBank mRNA | AK095693.1 | - | 5 | 2201 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM182A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026713.1 | 5 | 2242 | |||
GenBank mRNA | AK095693.1 | - | 5 | 2201 |