siRNA Details
Gene Symbol: FAM182A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026713.1 | 2, 3 | 765 | |||
GenBank mRNA | AK095693.1 | - | 2, 3 | 724 | ||
BC131537.1 | - | 2, 3 | 717 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: FAM182A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026713.1 | 2, 3 | 765 | |||
GenBank mRNA | AK095693.1 | - | 2, 3 | 724 | ||
BC131537.1 | - | 2, 3 | 717 |