siRNA Details
Gene Symbol: LOC440461 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027283.1 | 1 | 1232 | |||
GenBank mRNA | BC080624.1 | - | 1 | 1056 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV721347.1 | - | 1 | 333 | Chr. 17 - 64777688 - 64778411 [-] on Build GRCh38 |
