siRNA Details
Gene Symbol: DHRS4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023921.2 | 2 | 1542 | |||
NR_023922.2 | 2 | 1283 | ||||
NR_023923.2 | 3 | 1251 | ||||
NR_023924.2 | 2 | 1342 | ||||
GenBank mRNA | BX247982.1 | - | 2 | 1012 | ||
BX647964.1 | - | 3 | 1218 | |||
HQ845197.1 | - | 3 | 1237 | |||
HQ845198.1 | - | 3 | 1237 | |||
HQ845199.1 | - | 2 | 1322 | |||
JQ979294.1 | - | 3 | 1237 | |||
KJ545484.1 | - | 2 | 1177 | |||
KJ574270.1 | - | 2 | 968 | |||
KJ574271.1 | - | 2 | 1101 | |||
KJ574272.1 | - | 2 | 1101 | |||
KJ574273.1 | - | 2 | 1261 | |||
KJ574274.1 | - | 2 | 1305 | |||
KJ599455.1 | - | 2 | 1035 | |||
KJ599456.1 | - | 2 | 1292 | |||
KJ599457.1 | - | 2 | 994 | |||
KJ599458.1 | - | 1 | 788 | |||
KJ599459.1 | - | 3 | 1210 | |||
KJ599460.1 | - | 2 | 1336 | |||
KJ599461.1 | - | 3 | 1220 |
