siRNA Details
Gene Symbol: DHRS4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023921.2 | 2 | 2438 | |||
NR_023922.2 | 2 | 2179 | ||||
NR_023923.2 | 3 | 2147 | ||||
NR_023924.2 | 2 | 2238 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: DHRS4-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_023921.2 | 2 | 2438 | |||
NR_023922.2 | 2 | 2179 | ||||
NR_023923.2 | 3 | 2147 | ||||
NR_023924.2 | 2 | 2238 |