siRNA Details
Gene Symbol: TMEM254-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027428.1 | 6 | 1352 | |||
NR_027429.1 | 5 | 1265 | ||||
NR_027430.1 | 5 | 1141 | ||||
NR_027431.2 | 5 | 1115 | ||||
NR_027432.1 | 4 | 1028 | ||||
GenBank mRNA | AK021673.1 | - | 5 | 984 | ||
AK096758.1 | - | 6 | 1054 | |||
AK308811.1 | - | 4 | 581 | |||
AK309654.1 | - | 5 | 764 | |||
AK310182.1 | - | 5 | 817 | |||
AK310261.1 | - | 4 | 718 | |||
BC028046.1 | - | 4 | 689 |
