siRNA Details
Gene Symbol: LOC100129055 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024524.1 | 1 | 84 | |||
GenBank mRNA | BC046442.1 | - | 1 | 87 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG721123.1 | - | 1 | 92 | Chr. 10 - 38175668 - 38212357 [+] on Build GRCh38 |
