siRNA Details
Gene Symbol: LOC100129055 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024524.1 | 7 | 2615 | |||
GenBank mRNA | BC046442.1 | - | 7 | 2616 |
Gene Symbol: CCNYL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_103829.1 | 9 | 3146 |
