siRNA Details
Gene Symbol: SMIM10L2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024359.2 | 2 | 3191 | |||
GenBank mRNA | CA389121.1 | - | 1 | 141 | ||
CN430413.1 | - | 1 | 495 | |||
DB051081.1 | - | 1 | 68 | |||
HY002174.1 | - | 1 | 279 |
