siRNA Details
Gene Symbol: SMIM10L2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024359.2 | 2 | 3378 | |||
GenBank mRNA | CA389121.1 | - | 1 | 328 | ||
DB051081.1 | - | 1 | 256 | |||
HY002174.1 | - | 1 | 466 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: SMIM10L2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024359.2 | 2 | 3378 | |||
GenBank mRNA | CA389121.1 | - | 1 | 328 | ||
DB051081.1 | - | 1 | 256 | |||
HY002174.1 | - | 1 | 466 |