siRNA Details
Gene Symbol: SMIM10L2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024359.2 | 2 | 3212 | |||
GenBank mRNA | CA389121.1 | - | 1 | 162 | ||
CN430413.1 | - | 1 | 516 | |||
DB051081.1 | - | 1 | 89 | |||
HY002174.1 | - | 1 | 300 |
