siRNA Details
Gene Symbol: LINC00200 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_015376.2 | 2, 3 | 316 | |||
GenBank mRNA | AK097673.1 | - | 2, 3 | 266 | ||
BC133038.1 | - | 2, 3 | 316 | |||
BC133040.1 | - | 2, 3 | 268 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB045405.1 | - | 2, 3 | 266 | Chr. 10 - 1159821 - 1162846 [+] on Build GRCh38 |
