siRNA Details
Gene Symbol: LOC285074 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026846.1 | 2, 3 | 640 | |||
GenBank mRNA | BC069048.1 | - | 2, 3 | 422 | ||
BC132791.1 | - | 2, 3 | 381 | |||
BC132793.1 | - | 2, 3 | 381 | |||
BM454994.1 | - | 2, 3 | 593 | |||
BX353812.2 | - | 2, 3 | 641 |
