siRNA Details
Gene Symbol: LOC285074 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026846.1 | 11 | 2022 | |||
GenBank mRNA | AK057663.1 | - | 7 | 2876 | ||
BC069048.1 | - | 11 | 1804 | |||
DQ786261.1 | - | 1 | 307 |
Gene Symbol: LOC730268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017005464.1 | XP_016860953.1 | 14 | 1828 |
