siRNA Details
Gene Symbol: LINC00924 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027132.1 | 3, 4 | 348 | |||
NR_027133.1 | 3, 4 | 348 | ||||
GenBank mRNA | AK057337.1 | - | 3, 4 | 348 | ||
BC131607.1 | - | 3, 4 | 347 | |||
DB021433.1 | - | 3, 4 | 348 |
