siRNA Details
Gene Symbol: DRAIC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026979.1 | 1 | 59 | |||
GenBank mRNA | BC029061.1 | - | 1 | 59 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI762341.1 | - | 1 | 58 | Chr. 15 - 69561723 - 69565363 [+] on Build GRCh38 |
